Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ULBP-4/RAET1E Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6285G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6285G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6285G100 Supplier R&D Systems Supplier No. FAB6285G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ULBP-4/RAET1E Monoclonal specifically detects ULBP-4/RAET1E in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ULBP-4/RAET1E
Applications Flow Cytometry
Classification Monoclonal
Clone 709116
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q8TD07
Gene Alias bA350J20.7, LETAL, LETALRAE-1-like transcript 4, Lymphocyte effector toxicity activation ligand, MGC125309, N2DL4, N2DL-4, NKG2D ligand 4, NKG2DL4, RAET1E, RAET1E2, retinoic acid early transcript 1ERL-4, RL-4, ULBP4, ULBP4MGC125308
Gene Symbols RAET1E
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ULBP-4/RAET1E Gly30-Asp225 Accession # Q8TD07
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135250
Test Specificity Detects human ULBP-4/RAET1E in direct ELISAs and Western blots. In Western blots, approximately 20% cross-reactivity with recombinant human (rh) ULBP-2 is observed and no cross-reactivity with rhULBP-1 or rhULBP-3 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.