Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ULBP-4/RAET1E Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Supplier: R&D Systems MAB6285
This item is not returnable.
View return policy
Description
ULBP-4/RAET1E Monoclonal specifically detects ULBP-4/RAET1E in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
ULBP-4/RAET1E | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready | |
Q8TD07 | |
RAET1E | |
Mouse myeloma cell line NS0-derived recombinant human ULBP-4/RAET1E Gly30-Asp225 Accession # Q8TD07 | |
100 μg | |
Primary | |
Detects human ULBP-4/RAET1E in direct ELISAs and Western blots. In Western blots, approximately 20% cross-reactivity with recombinant human (rh) ULBP-2 is observed and no cross-reactivity with rhULBP-1 or rhULBP-3 is observed. | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
709116 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
bA350J20.7, LETAL, LETALRAE-1-like transcript 4, Lymphocyte effector toxicity activation ligand, MGC125309, N2DL4, N2DL-4, NKG2D ligand 4, NKG2DL4, RAET1E, RAET1E2, retinoic acid early transcript 1ERL-4, RL-4, ULBP4, ULBP4MGC125308 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
135250 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction