Learn More
Human ULBP-4/RAET1E Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | ULBP-4/RAET1E |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 ug/mL |
| Gene Accession No. | Q8TD07 |
| Gene Alias | bA350J20.7, LETAL, LETALRAE-1-like transcript 4, Lymphocyte effector toxicity activation ligand, MGC125309, N2DL4, N2DL-4, NKG2D ligand 4, NKG2DL4, RAET1E, RAET1E2, retinoic acid early transcript 1ERL-4, RL-4, ULBP4, ULBP4MGC125308 |
| Gene Symbols | RAET1E |
| Host Species | Goat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.