Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human UNC13D Alternative Isoform Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8966
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8966 100 μg
MAB8966SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB8966 Supplier R&D Systems Supplier No. MAB8966
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

UNC13D/Munc 13-4 Monoclonal specifically detects UNC13D/Munc 13-4 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.

Specifications

Antigen UNC13D/Munc 13-4
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1222B
Conjugate Unconjugated
Dilution Western Blot 0.05 ug/mL, Simple Western 0.5 ug/mL, Immunocytochemistry 2-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Gene Accession No. EAW89322
Gene Alias FHL3, HLH3, HPLH3, MUNC13-4, Munc13-4protein unc-13 homolog D, unc-13 homolog D (C. elegans)
Gene Symbols UNC13D
Host Species Rabbit
Immunogen KLH-coupled N-terminal human UNC13D peptide of an alternative isoform described in PMID:24842371 Accession # EAW89322
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201294
Test Specificity Detects the alternate form of human UNC13D in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.