Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human UNC13D Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $468.00
Specifications
Antigen | UNC13D/Munc 13-4 |
---|---|
Clone | 1223A |
Dilution | Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunocytochemistry 5-25 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB89661SP
![]() |
R&D Systems
MAB89661SP |
25 μg |
Each for $200.50
|
|
|||||
MAB89661
![]() |
R&D Systems
MAB89661 |
100 μg |
Each for $468.00
|
|
|||||
Description
UNC13D/Munc 13-4 Monoclonal specifically detects UNC13D/Munc 13-4 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
UNC13D/Munc 13-4 | |
Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunocytochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
FHL3, HLH3, HPLH3, MUNC13-4, Munc13-4protein unc-13 homolog D, unc-13 homolog D (C. elegans) | |
UNC13D | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
1223A | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q70J99 | |
201294 | |
KLH-coupled N-terminal human UNC13D peptide Accession # Q70J99 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS | |
Detects the conventional form of human UNC13D in Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title