Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human UNC5H3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1005
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1005 500 μg
MAB1005SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1005 Supplier R&D Systems Supplier No. MAB1005
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

UNC5H3/UNC5C Monoclonal specifically detects UNC5H3/UNC5C in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen UNC5H3/UNC5C
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 200414
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O95185
Gene Alias netrin receptor UNC5C, Protein unc-5 homolog 3, Protein unc-5 homolog C, unc-5 homolog 3, unc-5 homolog C (C. elegans), UNC5C, UNC5H3unc5 (C.elegans homolog) c
Gene Symbols UNC5C
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human UNC5H3 Ala40-Asp376 Accession # O95185
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8633
Test Specificity Detects human UNC5H3 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human UNC5H4 or recombinant rat UNC5H2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.