Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human UNC5H4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1429
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1429 100 μg
AF1429SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF1429 Supplier R&D Systems Supplier No. AF1429
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 3 publications

UNC5H4/UNC5D Polyclonal specifically detects UNC5H4/UNC5D in Human samples. It is validated for Western Blot.

Specifications

Antigen UNC5H4/UNC5D
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. NP_543148
Gene Alias FLJ16019, KIAA1777Unc5h4, netrin receptor UNC5D, netrin receptor Unc5h4, PRO34692, Protein unc-5 homolog 4, Protein unc-5 homolog D, unc-5 homolog 4, unc-5 homolog D (C. elegans), UNC5D, UNC5H4
Gene Symbols UNC5D
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human UNC5H4 Gly15-Ser373 Accession # NP_543148
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 137970
Test Specificity Detects human UNC5H4 in direct ELISAs and Western blots. In these formats, less than 5% cross-reactivity with recombinant rat (rr) UNC5H1, rrUNC5H2, and recombinant human UNC5H3 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.