Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human URB Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3410SP
This item is not returnable.
View return policy
Description
URB/CCD80 Polyclonal specifically detects URB/CCD80 in Human samples. It is validated for Western Blot.Specifications
URB/CCD80 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CCDC80, coiled-coil domain containing 80, down-regulated by oncogenes protein 1, DRO1, SSG1, Steroid-sensitive protein 1 | |
Goat | |
Affinity Purified | |
RUO | |
151887 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Q76M96 | |
CCDC80 | |
Mouse myeloma cell line NS0-derived recombinant human URB Met129-Tyr950 Accession # Q76M96 | |
25 μg | |
Primary | |
Detects human URB in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction