Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Uromodulin Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5144SP
This item is not returnable.
View return policy
Description
Uromodulin Polyclonal specifically detects Uromodulin in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Uromodulin | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ADMCKD2, FJHN, HNFJ, HNFJ1, MCKD2, Tamm-Horsfall glycoprotein, Tamm-Horsfall urinary glycoprotein, THGP, THP, UMOD, uromodulin, uromodulin (uromucoid, Tamm-Horsfall glycoprotein), uromucoid | |
Sheep | |
Affinity Purified | |
RUO | |
7369 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunohistochemistry 2-15 ug/mL | |
P07911 | |
UMOD | |
Mouse myeloma cell line NS0-derived recombinant human Uromodulin isoform 1 Asp25-Ser614 Accession # P07911 | |
25 μg | |
Primary | |
Detects human Uromodulin in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant mouse Uromodulin is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction