Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Uteroglobin/SCGB1A1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4218SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4218SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4218SP Supplier R&D Systems Supplier No. MAB4218SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 2 publications

Uteroglobin/SCGB1A1 Monoclonal specifically detects Uteroglobin/SCGB1A1 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Uteroglobin/SCGB1A1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 394324
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P11684
Gene Alias blastokinin, CC10, CC10Urine protein 1, CC16, CCPBP, CCSP, CCSPCCPBP, Clara, Clara cell phospholipid-binding protein, Clara cells 10 kDa secretory protein, PCB-BP, SCGB1A1, Secretoglobin family 1A member 1, secretoglobin, family 1A, member 1 (uteroglobin), UGB, UGBUP-1, UP1, Urinary protein 1, uteroglobin
Gene Symbols SCGB1A1
Host Species Rat
Immunogen E. coli-derived recombinant human Uteroglobin/SCGB1A1 Glu22-Asn91 Accession # P11684
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7356
Test Specificity Detects human Uteroglobin/SCGB1A1 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.