Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Uteroglobin/SCGB1A1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4218
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4218 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4218 Supplier R&D Systems Supplier No. AF4218
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Uteroglobin/SCGB1A1 Polyclonal specifically detects Uteroglobin/SCGB1A1 in Human samples. It is validated for Immunohistochemistry, Neutralization.

Specifications

Antigen Uteroglobin/SCGB1A1
Applications Immunohistochemistry, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 5-15 ug/mL, Neutralization 0.3-1.2 ug/mL
Gene Accession No. NP_003348
Gene Alias blastokinin, CC10, CC10Urine protein 1, CC16, CCPBP, CCSP, CCSPCCPBP, Clara, Clara cell phospholipid-binding protein, Clara cells 10 kDa secretory protein, PCB-BP, SCGB1A1, Secretoglobin family 1A member 1, secretoglobin, family 1A, member 1 (uteroglobin), UGB, UGBUP-1, UP1, Urinary protein 1, uteroglobin
Gene Symbols SCGB1A1
Host Species Sheep
Immunogen E. coli-derived recombinant human Uteroglobin/SCGB1A1 Glu22-Asn91 Accession # NP_003348
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7356
Test Specificity Detects human Uteroglobin/SCGB1A1 in direct ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.