Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human UVRAG Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC9180R025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC9180R025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC9180R025 Supplier R&D Systems Supplier No. IC9180R025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

UVRAG Monoclonal specifically detects UVRAG in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen UVRAG
Applications Flow Cytometry
Classification Monoclonal
Clone 953035
Conjugate Alexa Fluor 647
Dilution Intracellular Staining by Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q9P2Y5
Gene Alias beclin 1 binding protein, DHTX, disrupted in heterotaxy, p63, UV radiation resistance associated gene, UV radiation resistance-associated gene protein, VPS38
Host Species Mouse
Immunogen Human UVRAG synthetic peptide Accession # Q9P2Y5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7405
Test Specificity Detects human UVRAG in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.