Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human UVRAG Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9180SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9180SP 25 μg
MAB9180100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9180SP Supplier R&D Systems Supplier No. MAB9180SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

UVRAG Monoclonal specifically detects UVRAG in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen UVRAG
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 953035
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9P2Y5
Gene Alias beclin 1 binding protein, DHTX, disrupted in heterotaxy, p63, UV radiation resistance associated gene, UV radiation resistance-associated gene protein, VPS38
Host Species Mouse
Immunogen Human UVRAG synthetic peptide corresponding to sequences between amino acids 260-360 Accession # Q9P2Y5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7405
Test Specificity Detects human UVRAG in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.