Learn More
Human V1a Vasopressin R/AVPR1A Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | V1a Vasopressin R/AVPR1A |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 612130 |
| Conjugate | Alexa Fluor 405 |
| Dilution | Flow Cytometry |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide |
| Gene Accession No. | P37288 |
| Gene Alias | Antidiuretic hormone receptor 1a, arginine vasopressin receptor 1A, AVPR V1a, AVPR1, AVPR1A, SCCL vasopressin subtype 1a receptor, V1a vasopressin receptor, V1a VasopressinR, V1aR, V1-vascular vasopressin receptor AVPR1A, Vascular/hepatic-type arginine vasopressin receptor, vasopressin V1a receptor |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.