Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human V1a Vasopressin R/AVPR1A Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10932SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10932SP 25 μg
MAB10932100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10932SP Supplier R&D Systems Supplier No. MAB10932SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

V1a Vasopressin R/AVPR1A Monoclonal antibody specifically detects V1a Vasopressin R/AVPR1A in Human samples. It is validated for Flow Cytometry

Specifications

Antigen V1a Vasopressin R/AVPR1A
Applications Flow Cytometry
Classification Monoclonal
Clone 612130
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene Accession No. P37288
Gene Alias Antidiuretic hormone receptor 1a, arginine vasopressin receptor 1A, AVPR V1a, AVPR1, AVPR1A, SCCL vasopressin subtype 1a receptor, V1a vasopressin receptor, V1a VasopressinR, V1aR, V1-vascular vasopressin receptor AVPR1A, Vascular/hepatic-type arginine vasopressin receptor, vasopressin V1a receptor
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human V1a Vasopressin R/AVPR1A, Accession # P37288
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 552
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.