Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VAMP-8 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF5354SP
This item is not returnable.
View return policy
Description
VAMP-8 Polyclonal specifically detects VAMP-8 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
VAMP-8 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EDB, EDBVAMP-8, Endobrevin, VAMP8, vesicle-associated membrane protein 8, vesicle-associated membrane protein 8 (endobrevin) | |
Goat | |
Affinity Purified | |
RUO | |
8673 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 1-15 ug/mL | |
Q9BV40 | |
VAMP8 | |
E. coli-derived recombinant human VAMP-8 Met1-Lys75 Accession # Q9BV40 | |
25 μg | |
Primary | |
Detects human VAMP-8 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) VAMP-1, rhVAMP-2, rhVAMP-5, and rhVAMP-7 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction