Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Vang-like Protein 1/VANGL1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5476
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5476 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5476 Supplier R&D Systems Supplier No. MAB5476
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Vang-like Protein 1/VANGL1 Monoclonal specifically detects Vang-like Protein 1/VANGL1 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen Vang-like Protein 1/VANGL1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 497707
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8TAA9
Gene Alias KAI1 C-terminal interacting tetraspanin, Loop-tail protein 2 homolog, LPP2, MGC5338, STB2, STB2KITENIN, STBM2, Strabismus 2, Van Gogh-like protein 1, vang (van gogh, Drosophila)-like 1, vang, van gogh-like 1 (Drosophila), Vang like Protein 1, VANGL1, vang-like 1 (van gogh, Drosophila), vang-like protein 1
Gene Symbols VANGL1
Host Species Mouse
Immunogen E. coli-derived recombinant human Vang-like Protein 1/VANGL1 Glu242-Val524 Accession # Q8TAA9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81839
Test Specificity Detects human Vang-like Protein 1/VANGL1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, slight cross-reactivity with recombinant human VANGL2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.