Learn More
Description
Specifications
Specifications
| Assay Range | 31.3 - 2,000pg/mL |
| Assay Sensitivity | 19pg/mL |
| Product Type | ELISA Kit |
| Sample Type | Amniotic Fluid, Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 8639 |
| Gene Symbol | AOC3 |
| Kit Contents | Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human sVAP-1, Biotin-Conjugate anti-human sVAP-1 monoclonal antibody, Streptavidin-HRP, Human sVAP-1 Standard lyophilized, 4ng/mL upon reconstitution, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films |
| Quantity | 96 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
