Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VAP-B Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5855
This item is not returnable.
View return policy
Description
VAP-B Polyclonal specifically detects VAP-B in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
VAP-B | |
Polyclonal | |
Unconjugated | |
O95292 | |
VAPB | |
E. coli-derived recombinant human VAP-B Ala2-Pro132 Accession # O95292 | |
100 μg | |
Primary | |
Detects human VAP-B in direct ELISAs and Western blots. In direct ELISAs, approximately 60% cross-reactivity with recombinant rat VAP-B is observed and less than 5% cross-reactivity with recombinant human VAP-A is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
ALS8, ALS8VAMP-B/VAMP-C, VAMP (vesicle-associated membrane protein)-associated protein B and C, VAMP-associated 33 kDa protein, VAMP-associated protein B/C, VAMP-B, VAMP-C, VAPB, VAP-B, VAP-B/VAP-C, VAPC, VAP-C, vesicle-associated membrane protein-associated protein B/C | |
Sheep | |
Affinity Purified | |
RUO | |
9217 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction