Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Vasohibin-1 ELISA Kit

Catalog No. EH480RBX10
Encompass_Preferred
Click to view available options
Quantity:
10 x 96 Tests

ELISA

VASH1 is a protein that possesses actin binding activity and metallocarboxypeptidase activity. It is involved in several processes including negative regulation of angiogenesis, negative regulation of blood vessel endothelial cell migration, and proteolysis. VASH1 acts upstream of or within processes such as negative regulation of endothelial cell migration, negative regulation of endothelial cell proliferation, and negative regulation of lymphangiogenesis. It is located in the apical part of the cell, endoplasmic reticulum, and extracellular space. VASH1 is implicated in liver cirrhosis and portal hypertension, and is a biomarker of liver cirrhosis. Diseases associated with VASH1 include Portal Hypertension and Myasthenic Syndrome, Congenital, 14.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q7L8A9
Assay Range 11 to 2,500 pg/mL
Assay Sensitivity 11 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 22846
Gene Symbol VASH1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target VASH1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.