Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Vasorin/SLIT-like 2 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $468.50
Specifications
Antigen | Vasorin/SLIT-like 2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Vasorin/SLIT-like 2 Polyclonal specifically detects Vasorin/SLIT-like 2 in Human samples. It is validated for Western Blot.Specifications
Vasorin/SLIT-like 2 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
114990 | |
Chinese hamster ovary cell line CHO-derived recombinant human Vasorin extracellular domain | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Vasorin in direct ELISAs and Western blots. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Atia, Protein slit-like 2, SLIT like 2, SLITL2, slit-like 2, slit-like 2 (Drosophila), VASN, vasorin | |
VASN | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title