Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Vasorin/SLIT-like 2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2140
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2140 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB2140 Supplier R&D Systems Supplier No. MAB2140
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

Vasorin/SLIT-like 2 Monoclonal specifically detects Vasorin/SLIT-like 2 in Human samples. It is validated for Western Blot.

Specifications

Antigen Vasorin/SLIT-like 2
Applications Western Blot
Classification Monoclonal
Clone 227526
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q6EMK4
Gene Alias Atia, Protein slit-like 2, SLIT like 2, SLITL2, slit-like 2, slit-like 2 (Drosophila), VASN, vasorin
Gene Symbols VASN
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Vasorin aa 24-575 Accession # Q6EMK4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114990
Test Specificity Detects human Vasorin in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.