Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VCAM-1/CD106 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 6 publications
Supplier: R&D Systems BBA19
This item is not returnable.
View return policy
Description
VCAM-1/CD106 Polyclonal specifically detects VCAM-1/CD106 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
VCAM-1/CD106 | |
Polyclonal | |
Unconjugated | |
CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1 | |
Goat | |
500 μL | |
Primary | |
Detects VCAM-1/CD106 in Western blots. VCAM-1/CD106 has been screened using CHO cells transfected with cDNAs for ICAM-1 and VCAM-1. | |
Human | |
Serum |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1:1000 dilution, Simple Western 20 ug/mL, Immunohistochemistry 1:200 dilution | |
VCAM1 | |
Chinese hamster ovary cell line CHO-derived recombinant human VCAM-1 | |
RUO | |
7412 | |
Reconstitute in 0.5 mL of sterile water. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction