Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VCAM-1/CD106 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 33 publications
$552.44
Specifications
| Antigen | VCAM-1/CD106 |
|---|---|
| Clone | BBIG-V1 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
VCAM-1/CD106 Monoclonal specifically detects VCAM-1/CD106 in Human samples. It is validated for Western Blot, Immunoprecipitation, Adhesion Blockade, Immunocytochemistry.Specifications
| VCAM-1/CD106 | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant |
| BBIG-V1 | |
| Unconjugated | |
| Mouse | |
| Human | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title