Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VCAM-1/CD106 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $467.50
Specifications
Antigen | VCAM-1/CD106 |
---|---|
Clone | 1027433 |
Dilution | Western Blot 2 ug/mL, Simple Western 20 ug/mL, Flow Cytometry 0.25 ug/106 cells, Immunohistochemistry 5-25 ug/mL, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB8091SP
![]() |
R&D Systems
MAB8091SP |
25 μg |
Each for $200.50
|
|
|||||
MAB8091100
![]() |
R&D Systems
MAB8091100 |
100 μg |
Each for $467.50
|
|
|||||
Description
VCAM-1/CD106 Monoclonal antibody specifically detects VCAM-1/CD106 in Human samples. It is validated for Western Blot, Western Blot, Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
VCAM-1/CD106 | |
Western Blot 2 ug/mL, Simple Western 20 ug/mL, Flow Cytometry 0.25 ug/106 cells, Immunohistochemistry 5-25 ug/mL, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1 | |
Chinese Hamster Ovary cell line CHO-derived human VCAM-1/CD106, Phe25-Glu698, Accession # P19320-1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1027433 | |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
P19320-1 | |
7412 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title