Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human VEGF Alexa Fluor 488-conjugated Antibody, R&D Systems™

Catalog No. IC2931G
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC2931G 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC2931G Supplier R&D Systems Supplier No. IC2931G
This item is not returnable. View return policy

Mouse Monoclonal Antibody

VEGF Monoclonal specifically detects VEGF in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen VEGF
Applications Flow Cytometry
Classification Monoclonal
Clone 23410
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Alias MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor
Gene Symbols VEGFA
Host Species Mouse
Immunogen E. coli-derived recombinant human VEGF
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7422
Test Specificity Detects human VEGF in flow cytometry.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.