Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VEGF-B167/186 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
$200.50 - $573.00
Specifications
Antigen | VEGF-B |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
VEGF-B Polyclonal specifically detects VEGF-B in Human samples. It is validated for Western Blot.Specifications
VEGF-B | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
7423 | |
E. coli-derived recombinant human VEGF-B 186 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human VEGF-B 167 and human VEGF-B 186 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) VEGF-B 186, rmVEGF-B 167, rmVEGF-B 206, recombinant human (rh) VEGF-A, rhVEGF-C, and rhVEGF-D is observed. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
vascular endothelial growth factor B, VEGFB, VEGF-B, VEGF-related factor, VRFVEGFL | |
VEGFB | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title