Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VEGF-B186 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF337
This item is not returnable.
View return policy
Description
VEGF-B Polyclonal specifically detects VEGF-B in Human samples. It is validated for Western Blot.Specifications
VEGF-B | |
Polyclonal | |
Unconjugated | |
P49765 | |
VEGFB | |
E. coli-derived recombinant human VEGF-B 186 Accession # P49765 | |
100 μg | |
Primary | |
Detects human VEGF-B 186 in direct ELISAs and Western blots. In these formats, this antibody shows 30% cross-reactivity with rmVEGF-B 186 and less than 1% cross-reactivity with rhVEGF-B 167 rhVEGF-D, and rmVEGF-D. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
vascular endothelial growth factor B, VEGFB, VEGF-B, VEGF-related factor, VRFVEGFL | |
Goat | |
Affinity Purified | |
RUO | |
7423 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction