Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human VEGF-C Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB752500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB752500 500 μg
MAB752SP 25 μg
MAB752100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB752500 Supplier R&D Systems Supplier No. MAB752500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

VEGF-C Monoclonal specifically detects VEGF-C in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen VEGF-C
Applications Western Blot, ELISA
Classification Monoclonal
Clone 193208
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q6FH59
Gene Alias Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGFC, VEGF-C, VRPFLT4 ligand DHM
Gene Symbols VEGFC
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human VEGF-C Thr103-Arg227 (Cys156Ser) Accession # Q6FH59
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7424
Test Specificity Detects human VEGF-C in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) PDGF-AA, rhPDGF-BB, rhPlGF, rhVEGF 165 rhVEGF 206 rhVEGF-B 186 or rhVEGF-D.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.