Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human VEGF-D Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 3 publications

$200.50 - $1973.00

Specifications

Antigen VEGF-D
Clone 78902
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB2861SP
GREENER_CHOICE
View Documents
R&D Systems
MAB2861SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB2861100
GREENER_CHOICE
View Documents
R&D Systems
MAB2861100
100 μg
Each for $538.00
Only null left
Add to Cart
 
MAB2861500
GREENER_CHOICE
View Documents
R&D Systems
MAB2861500
500 μg
Each for $1,973.00
Only null left
Add to Cart
 
Description

Description

VEGF-D Monoclonal specifically detects VEGF-D in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications

Specifications

VEGF-D
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Monoclonal
Mouse
Human
O43915
2277
Mouse myeloma cell line NS0-derived recombinant human VEGF-D Phe93-Ser201 Accession # O43915
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human VEGF-D in ELISAs and Western blots. In ELISAs, this antibody shows less than 1% cross-reactivity with recombinant human (rh) VEGF 121 rhVEGF 165 rhVEGF-B 167 rhVEGF-B 186 and rhVEGF-C. In Western blots, this antibody shows 100% cross-reactivity with rmVEGF-D, less than 5% cross-reactivity with rhVEGF 121 rhVEGF 165 rmVEGF 120 and no cross-reactivity with rhVEGF-B 165 rmVEGF-B 186 rhVEGF-B 186 rmVEGF 115 rrVEGF 164 or rhVEGF 206 .
78902
Western Blot, ELISA
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
c-fos induced growth factor (vascular endothelial growth factor D), FIGF, vascular endothelial growth factor D, VEGFD, VEGF-DVEGFDc-Fos-induced growth factor
FIGF
IgG1
Protein A or G purified from ascites
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.