Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human VEGF-D Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB286100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB286100 100 μg
MAB286SP 25 μg
MAB286500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB286100 Supplier R&D Systems Supplier No. MAB286100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

VEGF-D Monoclonal specifically detects VEGF-D in Human samples. It is validated for Immunohistochemistry, Neutralization.

Specifications

Antigen VEGF-D
Applications Immunohistochemistry, Neutralization
Classification Monoclonal
Clone 78923
Conjugate Unconjugated
Dilution Immunohistochemistry 1-15 ug/mL, Neutralization 3-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O43915
Gene Alias c-fos induced growth factor (vascular endothelial growth factor D), FIGF, vascular endothelial growth factor D, VEGFD, VEGF-DVEGFDc-Fos-induced growth factor
Gene Symbols FIGF
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human VEGF-D Phe93-Ser201 Accession # O43915
Purification Method Protein A or G purified from ascites
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2277
Test Specificity Detects human VEGF-D in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) VEGF-D, rmVEGF-B, recombinant human (rh) VEGF-B and rhVEGF is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Ascites
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.