Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VEGF-D Biotinylated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems BAM286
This item is not returnable.
View return policy
Description
VEGF-D Monoclonal specifically detects VEGF-D in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
VEGF-D | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.5-2.0 ug/mL | |
O43915 | |
FIGF | |
Mouse myeloma cell line NS0-derived recombinant human VEGF-D Phe93-Ser201 Accession # O43915 | |
250 μg | |
Primary | |
Detects human VEGF-D in ELISAs and Western blots. In Western blots, this antibody shows approximately 2% cross-reactivity with recombinant mouse (rm) VEGF-D and no cross-reactivity with rhVEGF 165 rmVEGF 165 rhVEGF-B 186 rmVEGF-B 186 rhVEGF 206 rhVEGF-C, rhCTGF, or rhPlGF. | |
Human | |
Purified |
Western Blot, ELISA | |
78923 | |
Biotin | |
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
c-fos induced growth factor (vascular endothelial growth factor D), FIGF, vascular endothelial growth factor D, VEGFD, VEGF-DVEGFDc-Fos-induced growth factor | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
2277 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction