Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human VEGF/PlGF Heterodimer Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB297
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB297 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB297 Supplier R&D Systems Supplier No. MAB297
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

VEGF/PlGF Heterodimer Monoclonal specifically detects VEGF/PlGF Heterodimer in Human samples. It is validated for Western Blot.

Specifications

Antigen VEGF/PlGF Heterodimer
Applications Western Blot
Classification Monoclonal
Clone 146213
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAM03108 (VEGF) & AAD30179 (PIGF)
Host Species Mouse
Immunogen E. coli-derived recombinant human VEGF/PlGF heterodimer Ala27-Arg191 (VEGF) & Ala21-Arg149 (PIGF) Accession # AAM03108 (VEGF) & AAD30179 (PIGF)
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects human VEGF/PlGF heterodimer in direct ELISAs and Western blots. This antibody does not detect either subunit alone. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) VEGF 121 rhVEGF 165 rhVEGF 206 rhCTGF, rhLDGF, rhPlGF, rhVEGF-B 186 rhVEGF-C, rhVEGF-D, rhPDGF, rmVEGF 115 rmVEGF 120 rmVEGF 164 rmPlGF-2, rrVEGF 164 or rzfVEGF.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.