Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human VEGF Quantikine ELISA Kit

Catalog No. SVE00
Change view
Click to view available options
Quantity:
1 Plate
6 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SVE00 6 Plates
DVE00 1 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SVE00 Supplier R&D Systems™ Supplier No. SVE00
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human VEGF Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human VEGF Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human VEGF in cell culture supernates (200 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul).

Specifications

Assay Range 15.60 to 1000 pg/mL (Cell Culture Supernates), 31.30 to 2000 pg/mL (Serum, Heparin Plasma, Citrate Plasma, EDTA Plasma)
Assay Sensitivity 5 pg/mL (Cell Culture Supernates), 9 pg/mL (Serum, Heparin Plasma, Citrate Plasma, EDTA Plasma)
Conjugate HRP
Cross Reactivity Cross-species reactivity observed with 1 or more species tested. <0.5% cross-reactivity observed with available related molecules.
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Specificity Natural and Recombinant Human VEGF. This assay also recognizes Recombinant Human VEGF165b.
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 7422
Quantity 6 Plates
Sample Volume Cell Culture Supernates (200uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL),Citrate Plasma (100uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.