Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VEGF165 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 9 publications
Supplier: R&D Systems AB293NA
This item is not returnable.
View return policy
Description
VEGF Polyclonal specifically detects VEGF in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Specifications
VEGF | |
Polyclonal | |
Unconjugated | |
AAV38412 | |
VEGFA | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human VEGF 165 Ala27-Arg191 Accession # AAV38412 | |
1 mg | |
Primary | |
Detects human VEGF 165 and human VEGF 121 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse VEGF and recombinant rat VEGF is observed. | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Neutralization | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.6-3.0 ug/mL | |
MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor | |
Goat | |
Protein A or G purified | |
RUO | |
7422 | |
Reconstitute at 1 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction