Learn More
Human VEGFR Phycoerythrin Antibody Sampler, R&D Systems™

Description
Specifications
Specifications
| Antigen | VEGFR1/R2/R3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 89106, 49560, 54733 |
| Conjugate | R-PE |
| Dilution | Flow Cytometry |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Host Species | Mouse |
| Quantity | 1 Kit |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.