Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VEGFR1/Flt-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB321SP
This item is not returnable.
View return policy
Description
VEGFR1/Flt-1 Monoclonal specifically detects VEGFR1/Flt-1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
VEGFR1/Flt-1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.10, EC 2.7.10.1, FLT, FLT1, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGF R1, VEGFR1, VEGFR-1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2321 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Flow Cytometry, CyTOF | |
49560 | |
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
NP_001153392 | |
FLT1 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human VEGFR1/Flt-1, Ser27-His687, Accession # NP_001153392 | |
25 μg | |
Primary | |
Detects human VEGFR1/Flt-1 in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse VEGFR1, recombinant human (rh) VEGFR2, rhVEGFR3, or rhVEGFR4. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction