Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VEGFR1/Flt-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$573.00
Specifications
Clone | 49560 |
---|---|
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Purified |
Host Species | Mouse |
Description
VEGFR1/Flt-1 Monoclonal specifically detects VEGFR1/Flt-1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
49560 | |
Unconjugated | |
Mouse | |
Human | |
Primary |
Monoclonal | |
Purified | |
RUO | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title