Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human VEGFR1/Flt-1 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB321P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB321P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB321P Supplier R&D Systems Supplier No. FAB321P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

VEGFR1/Flt-1 Monoclonal specifically detects VEGFR1/Flt-1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen VEGFR1/Flt-1
Applications Flow Cytometry
Classification Monoclonal
Clone 49560
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. NP_001153392
Gene Alias EC 2.7.10, EC 2.7.10.1, FLT, FLT1, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGF R1, VEGFR1, VEGFR-1
Gene Symbols FLT1
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human VEGF R1/Flt-1 Ser27-His687 Accession # NP_001153392
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2321
Test Specificity Detects human VEGF R1/Flt-1 in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse VEGF R1, recombinant human (rh) VEGF R2, rhVEGF R3, or rhVEGF R4.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.