Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VEGFR2/KDR/Flk-1 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB3572RU100UG
This item is not returnable.
View return policy
Description
VEGFR2/KDR/Flk-1 Monoclonal specifically detects VEGFR2/KDR/Flk-1 in Human samples. It is validated for Flow Cytometry.Specifications
VEGFR2/KDR/Flk-1 | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, Flk1, Flk-1, FLK1tyrosine kinase growth factor receptor, KDR, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), KRD1, Ly73, Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGF R2, VEGFR, VEGFR2, VEGFR-2 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human VEGFR2/KDR/Flk-1, Ala20-Glu764, Accession # P35968 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
89106R | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P35968 | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
3791 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction