Learn More
Human VEGFR2/KDR/Flk-1 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | VEGFR2/KDR/Flk-1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 89106 |
| Conjugate | Alexa Fluor 700 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Gene Accession No. | P35968 |
| Gene Alias | CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, Flk1, Flk-1, FLK1tyrosine kinase growth factor receptor, KDR, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), KRD1, Ly73, Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGF R2, VEGFR, VEGFR2, VEGFR-2 |
| Gene Symbols | KDR |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.