Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VEGFR3/Flt-4 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 6 publications
Specifications
| Antigen | VEGFR3/Flt-4 |
|---|---|
| Clone | 54703 |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
MAB3491SP
|
R&D Systems
MAB3491SP |
25 μg | N/A | N/A | N/A | ||||
|
MAB3491100
|
R&D Systems
MAB3491100 |
100 μg | N/A | N/A | N/A | ||||
|
MAB3491500
|
R&D Systems
MAB3491500 |
500 μg | N/A | N/A | N/A | ||||
Description
VEGFR3/Flt-4 Monoclonal specifically detects VEGFR3/Flt-4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
| VEGFR3/Flt-4 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P35916 | |
| 2324 | |
| Mouse myeloma cell line NS0-derived recombinant human VEGFR3/Flt-4, Tyr25-Ile776, Accession # P35916 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human VEGFR3/Flt-4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25-30% cross-reactivity with recombinant mouse VEGFR3 is observed and no cross-reactivity with recombinant human (rh) VEGFR1 or rhVEGFR2 is observed. |
| 54703 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.10, EC 2.7.10.1, Flt-4, FLT4, fms-related tyrosine kinase 4, LMPH1A, PCLFLT41, soluble VEGFR3 variant 1, soluble VEGFR3 variant 2, soluble VEGFR3 variant 3, Tyrosine-protein kinase receptor FLT4, vascular endothelial growth factor receptor 3, VEGF R3, VEGFR-3, VEGFR3Fms-like tyrosine kinase 4 | |
| FLT4 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title