Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human VEGFR3 (Flt-4) Fc Chimera Recombinant Protein

Catalog No. PIRP88179
Encompass
Click to view available options
Quantity:
1 mg
100 μg

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (900 μL) be added to the vial to prepare a stock solution of 1.11 mg/mL. Concentration is measured by UV.

Vascular endothelial growth factor is a key regulator of blood vessel development in embryos and angiogenesis in adult tissues. Unlike VEGF, the related VEGFC stimulates the growth of lymphatic vessels through its specificl ymphatic endothelial receptor VEGFR-3. In 1998, Dumont showed that targeted inactivation of the VEGFR-3 gene in mice resulted in defective blood vessel development in early embryos. Vasculogenesis and angiogenesis occurred, but large vessels became abnormally organized with defective lumens, leading to fluid accumulation in the pericardial cavity and cardiovascular failure at embryonic day 9.5. Thus, VEGFR3 has an essential role in the development of theembryonic cardiovascular system before the emergence of the lymphatic vessels.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P35916
For Use With (Application) ELISA
Format Lyophilized
Formulation PBS with 0.01% mannitol, 0.01% Tween 80, 5-8% trehalose and no preservative; pH 7.4
Gene ID (Entrez) 2324
Molecular Weight (g/mol) 111.3 kDa
Name Human VEGFR3 (Flt-4) Fc Chimera
Purification Method SDS-PAGE
Quantity 100 μg
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias AI323512; Chy; chylous ascites; Flt4; FLT-4; FLT41; fms related tyrosine kinase 4; FMS-like tyrosine kinase 4; fms-related tyrosine kinase 4; LMPH1A; PCL; receptor protein tyrosine kinase; soluble VEGFR 3; sVEGFR 3; tyrosine-protein kinase receptor FLT4; Vascular endothelial growth factor receptor 3; vascular endothelial growth factor receptor-3; VEGF R3; VEGFR3; VEGFR-3
Common Name VEGF Receptor 3
Gene Symbol FLT4
Biological Activity Measured by its binding ability in a functional ELISA. 1. Immobilized recombinant human VEGFR3 at 5 μg/ml (100 μL/well) can bind recombinant human VEGF-D at a linear range of 62.5-2000 ng/mL 2. Immobilized recombinant human VEGF-C at 10 μg/ml (100 μL/well) can bind recombinant human VEGFR3 (at a linear range of 0.64-80 ng/mL 3. Scatchard analysis showed the affinity constant (Kd) of recombinant human VEGF-C bound to recombinant human VEGFR3 was 1.4 nM.)
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence Human VEGFR3, amino acids Met1-Ile776 (Accession # NP_002011.2) with a C-terminal human IgG1 Fc region
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Activity Measured by its binding ability in a functional ELISA. 1. Immobilized recombinant human VEGFR3 at 5 μg/ml (100 μL/well) can bind recombinant human VEGF-D at a linear range of 62.5-2000 ng/mL 2. Immobilized recombinant human VEGF-C (at 10 μg/ml (100 μL/well) can bind recombinant human VEGFR3 (at a linear range of 0.64-80 ng/mL 3. Scatchard analysis showed the affinity constant (Kd) of recombinant human VEGF-C bound to recombinant human VEGFR3 was 1.4 nM.))
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade >97% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.