Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VHL Antibody, R&D Systems™

Mouse Monoclonal Antibody
$208.00 - $484.00
Specifications
Antigen | Von Hippel Lindau |
---|---|
Clone | 1066402 |
Dilution | Western Blot 2 μg/mL, Simple Western 20 μg/mL |
Applications | Western Blot, Simple Western |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11435SP
![]() |
R&D Systems
MAB11435SP |
25 μg |
Each for $208.00
|
|
|||||
MAB11435100
![]() |
R&D Systems
MAB11435100 |
100 μg |
Each for $484.00
|
|
|||||
Description
Von Hippel Lindau Monoclonal antibody specifically detects Von Hippel Lindau in Human samples. It is validated for Western Blot, Simple WesternSpecifications
Von Hippel Lindau | |
Western Blot 2 μg/mL, Simple Western 20 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
elongin binding protein, HRCA1, Protein G7, pVHL, RCA1, VHL1, von Hippel-Lindau disease tumor suppressor, von Hippel-Lindau syndrome, von Hippel-Lindau tumor suppressor | |
E. coli -derived recombinant human VHL, Asn150-Asp213, Accession # P40337 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1066402 | |
Western Blot, Simple Western | |
Unconjugated | |
Mouse | |
Human | |
P40337 | |
7428 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title