Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human VIGR/GPR126 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB10577G10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB10577G10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB10577G10 Supplier R&D Systems Supplier No. FAB10577G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

VIGR/GPR126 Monoclonal antibody specifically detects VIGR/GPR126 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen VIGR/GPR126
Applications Flow Cytometry
Classification Monoclonal
Clone 1044430
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. AAH75798.1
Gene Alias APG1, Developmentally regulated G-protein-coupled receptor, DREG, FLJ14937, G protein-coupled receptor 126, Gm222, GPR126, G-protein coupled receptor 126, HBV PreS1-transactivated protein 2, PS1TP2, Vascular inducible G protein-coupled receptor, vascular-inducible G protein-coupled receptor, VIGRPS1TP2
Host Species Mouse
Immunogen Human embryonic kidney cell HEK293-derived human VIGR/GPR126 protein, Cys38-Lys437, Accession # AAH75798.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57211
Reconstitution Reconstitution default not known
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.