Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human VIPR2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5416
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5416 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5416 Supplier R&D Systems Supplier No. MAB5416
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

VIPR2/VPAC2 Monoclonal specifically detects VIPR2/VPAC2 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen VIPR2/VPAC2
Applications Immunohistochemistry
Classification Monoclonal
Clone 476031
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P41587
Gene Alias FLJ16511, Helodermin-preferring VIP receptor, PACAP type III receptor, PACAPR3, PACAP-R3, PACAP-R-3, Pituitary adenylate cyclase-activating polypeptide type III receptor, vasoactive intestinal peptide receptor 2, vasoactive intestinal polypeptide receptor 2, VIP R2, VIP2R, VIP-R-2, VPAC2, VPCAP2R
Gene Symbols VIPR2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human VIP R2 Glu24-Ile438 Accession # P41587
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7434
Test Specificity Detects human VIP R2 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.