Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human VIPR2 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB5416P025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5416P025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB5416P025 Supplier R&D Systems Supplier No. FAB5416P025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

VIPR2/VPAC2 Monoclonal specifically detects VIPR2/VPAC2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen VIPR2/VPAC2
Applications Flow Cytometry
Classification Monoclonal
Clone 476031
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P41587
Gene Alias FLJ16511, Helodermin-preferring VIP receptor, PACAP type III receptor, PACAPR3, PACAP-R3, PACAP-R-3, Pituitary adenylate cyclase-activating polypeptide type III receptor, vasoactive intestinal peptide receptor 2, vasoactive intestinal polypeptide receptor 2, VIP R2, VIP2R, VIP-R-2, VPAC2, VPCAP2R
Gene Symbols VIPR2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human VIP R2 Glu24-Ile438 Accession # P41587
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7434
Test Specificity Detects human VIP R2 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.