Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VISTA/B7-H5/PD-1H Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB71267U100UG
This item is not returnable.
View return policy
Description
VISTA/B7-H5/PD-1H Monoclonal specifically detects VISTA/B7-H5/PD-1H in Human samples. It is validated for Flow Cytometry.Specifications
VISTA/B7-H5/PD-1H | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
4632428N05Rik, B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, Dies1, Gi24, PD1H, PD-1H, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR | |
Mouse myeloma cell line NS0-derived recombinant human VISTA/B7-H5/PD-1H, Phe33-Ala194, Accession # Q9H7M9 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
1011484 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9H7M9 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
64115 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction