Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human VISTA/B7-H5/PD-1H Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. F71261RG100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F71261RG100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. F71261RG100 Supplier R&D Systems Supplier No. FAB71261RG100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

VISTA/B7-H5/PD-1H Monoclonal specifically detects VISTA/B7-H5/PD-1H in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen VISTA/B7-H5/PD-1H
Applications Flow Cytometry
Classification Monoclonal
Clone 730804R
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide.
Gene Accession No. Q9H7M9
Gene Alias 4632428N05Rik, B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, Dies1, Gi24, PD1H, PD-1H, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human VISTA/B7-H5/PD-1H, Phe33-Ala194, Accession No. Q9H7M9
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 64115.0
Test Specificity Detects human VISTA/B7-H5/PD-1H in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.