Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VISTA/B7-H5/PD-1H Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $444.50
Specifications
| Antigen | VISTA/B7-H5/PD-1H |
|---|---|
| Clone | 730835 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB71264SP
![]() |
R&D Systems
MAB71264SP |
25 μg |
Each for $200.50
|
|
|||||
MAB71264100
![]() |
R&D Systems
MAB71264100 |
100 μg |
Each for $444.50
|
|
|||||
Description
VISTA/B7-H5/PD-1H Monoclonal specifically detects VISTA/B7-H5/PD-1H in Human samples. It is validated for ELISA.Specifications
| VISTA/B7-H5/PD-1H | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| Q9H7M9 | |
| 64115 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 730835 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 4632428N05Rik, B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, Dies1, Gi24, PD1H, PD-1H, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR | |
| Mouse myeloma cell line NS0-derived recombinant human VISTA/B7-H5/PD-1H Met1-Ala194 Accession # Q9H7M9 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human VISTA/B7-H5/PD-1H in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title